Learn More
Cayman Chemical Lipid CL1, Purity: ≥95%, CAS: 1450888-71-7, 5 mg
SDP
Catalog No. 502916231
Encompass_Preferred
An ionizable cationic trialkyl lipid; has been used in the formation of LNPs for delivering mRNA in vivo; LNPs containing lipid CL1 and encapsulating mRNA encoding human EPO increase blood EPO protein levels in mice; LNPs containing lipid CL1 and encapsulating mRNA encoding a reporter protein increase reporter expression in the liver in mice; LNPs containing lipid CL1 and encapsulating mRNA encoding influenza A HA increase serum anti-HA IgG antibody titers in a mouse model of HA vaccination
Catalog No. 50-291-6231 Supplier Cayman Chemical Supplier No. 383205 MG
An ionizable cationic trialkyl lipid; has been used in the formation of LNPs for delivering mRNA in vivo; LNPs containing lipid CL1 and encapsulating mRNA encoding human EPO increase blood EPO protein levels in mice; LNPs containing lipid CL1 and encapsulating mRNA encoding a reporter protein increase reporter expression in the liver in mice; LNPs containing lipid CL1 and encapsulating mRNA encoding influenza A HA increase serum anti-HA IgG antibody titers in a mouse model of HA vaccination