Learn More
Cayman Chemical Lisinopril, Purity: ≥98%, CAS: 76547-98-3, 100 mg
SDP
Catalog No. 502968195
Encompass_Preferred
An ACE inhibitor (IC50 = 1.2 nM); reduces the formation of endothelin-1 and increases NO in human vascular endothelial cells at 0.1 nM; inhibits the pressor response to angiotensin I in anesthetized rats and dogs (ID50s = 2.3 and 6.5 µg/kg, respectively)
Catalog No. 50-296-8195 Supplier Cayman Chemical Supplier No. 16833100 MG
An ACE inhibitor (IC50 = 1.2 nM); reduces the formation of endothelin-1 and increases NO in human vascular endothelial cells at 0.1 nM; inhibits the pressor response to angiotensin I in anesthetized rats and dogs (ID50s = 2.3 and 6.5 µg/kg, respectively)