Learn More
Cayman Chemical Lixivaptan, Purity: ≥97%, CAS: 168079-32-1, 10 mg
SDP
Catalog No. 502986166
Encompass_Preferred
A nonpeptide antagonist of vasopressin V2 receptors (Ki = 2.3 nM)
Catalog No. 50-298-6166 Supplier Cayman Chemical Supplier No. 2151110 MG
A nonpeptide antagonist of vasopressin V2 receptors (Ki = 2.3 nM)