Learn More
Cayman Chemical Lodoxamide, Purity: ≥95%, CAS: 53882-12-5, 50 mg
SDP
Catalog No. 502933226
Encompass_Preferred
A potent agonist of GPR35 (EC50 = 1.61 nM); inhibits histamine release in isolated rat peritoneal mast cells (IC50s = 0.1-50 µM); inhibits A23187-induced calcium influx in isolated rat mast cells; reduces antigen-induced histamine release from rat conjunctival tissue by 46% in vitro when used at a concentration of 10 µg/ml; dose-dependently reduces the immediate hypersensitivity response in rat conjunctiva in vivo; reduces mast cell degranulation in rat conjunctiva
Catalog No. 50-293-3226 Supplier Cayman Chemical Supplier No. 2399450 MG
A potent agonist of GPR35 (EC50 = 1.61 nM); inhibits histamine release in isolated rat peritoneal mast cells (IC50s = 0.1-50 µM); inhibits A23187-induced calcium influx in isolated rat mast cells; reduces antigen-induced histamine release from rat conjunctival tissue by 46% in vitro when used at a concentration of 10 µg/ml; dose-dependently reduces the immediate hypersensitivity response in rat conjunctiva in vivo; reduces mast cell degranulation in rat conjunctiva