Learn More
Cayman Chemical Loratadine, Purity: ≥98%, CAS: 79794-75-5, 5 mg
SDP
Catalog No. 502970489
Encompass_Preferred
A non-sedating antihistamine that acts as a selective inverse agonist of peripheral histamine H1 receptors (Ki = 35 nM); inhibits the release of LTC4 (IC50 = 8 µM) and histamine (IC50 = 11 µM) from rodent mast cells and inhibits allergic bronchospasm in guinea pigs (ED50 = 0.40 mg/kg)
Catalog No. 50-297-0489 Supplier Cayman Chemical Supplier No. 156255 MG
A non-sedating antihistamine that acts as a selective inverse agonist of peripheral histamine H1 receptors (Ki = 35 nM); inhibits the release of LTC4 (IC50 = 8 µM) and histamine (IC50 = 11 µM) from rodent mast cells and inhibits allergic bronchospasm in guinea pigs (ED50 = 0.40 mg/kg)