Learn More
Cayman Chemical LOXO-101, Purity: ≥98%, CAS: 1223403-58-4, 10 mg
SDP
Catalog No. 502941593
Encompass_Preferred
A pan-Trk inhibitor (IC50s = 2-20 nM for TrkA, TrkB, and TrkC); selective for TrkA, -B, and -C over a panel of 226 kinases at 1 μM; inhibits the growth of CUTO-3.29, KM12, and MO-91 patient-derived cancer cell lines (IC50s = <100, <10, and <10 nM, respectively); reduces tumor growth in a KM12 mouse xenograft model at 60 and 200 mg/kg
Catalog No. 50-294-1593 Supplier Cayman Chemical Supplier No. 2705610 MG
A pan-Trk inhibitor (IC50s = 2-20 nM for TrkA, TrkB, and TrkC); selective for TrkA, -B, and -C over a panel of 226 kinases at 1 μM; inhibits the growth of CUTO-3.29, KM12, and MO-91 patient-derived cancer cell lines (IC50s = <100, <10, and <10 nM, respectively); reduces tumor growth in a KM12 mouse xenograft model at 60 and 200 mg/kg