Learn More
Cayman Chemical LSN2814617, Purity: ≥98%, CAS: 1313498-08-6, 25 mg
SDP
Catalog No. 502947639
Encompass_Preferred
A positive allosteric modulator of mGluR5 receptors; potentiates glutamate-induced calcium mobilization in MCB3901 cells expressing human mGluR5 (EC50 = 52 nM); increases rat hippocampal mGluR5 receptor occupancy (ED50 = 13 mg/kg); increases wakefulness and decreases NREM and REM sleep in rats at 0.3, 1, and 3 mg/kg
Catalog No. 50-294-7639 Supplier Cayman Chemical Supplier No. 3360725 MG
A positive allosteric modulator of mGluR5 receptors; potentiates glutamate-induced calcium mobilization in MCB3901 cells expressing human mGluR5 (EC50 = 52 nM); increases rat hippocampal mGluR5 receptor occupancy (ED50 = 13 mg/kg); increases wakefulness and decreases NREM and REM sleep in rats at 0.3, 1, and 3 mg/kg