Learn More
Cayman Chemical LT175, Purity: ≥98%, CAS: 862901-87-9, 5 mg
SDP
Catalog No. 502980035
Encompass_Preferred
A dual PPARα/γ ligand that displays partial agonism toward PPARγ (EC50s = 0.22, 0.26, and 0.48 µM for hPPARα, mPPARα, and hPPARγ, respectively); improves glucose homeostasis in diet-induced insulin resistant mice
Catalog No. 50-298-0035 Supplier Cayman Chemical Supplier No. 176365 MG
A dual PPARα/γ ligand that displays partial agonism toward PPARγ (EC50s = 0.22, 0.26, and 0.48 µM for hPPARα, mPPARα, and hPPARγ, respectively); improves glucose homeostasis in diet-induced insulin resistant mice