Learn More
Cayman Chemical LXR-623, Purity: ≥98%, CAS: 875787-07-8, 1 mg
SDP
Catalog No. 502980941
Encompass_Preferred
An LXR modulator that acts as a partial agonist at LXRα and a full agonist at LXRβ (IC50s = 179 and 24 nM, respectively); orally bioavailable and readily passes the BBB; anti-atherogenic in animal models of atherosclerosis
Catalog No. 50-298-0941 Supplier Cayman Chemical Supplier No. 211171 MG
An LXR modulator that acts as a partial agonist at LXRα and a full agonist at LXRβ (IC50s = 179 and 24 nM, respectively); orally bioavailable and readily passes the BBB; anti-atherogenic in animal models of atherosclerosis