Learn More
Cayman Chemical LY2606368, Purity: ≥98%, CAS: 1234015-52-1, 10 mg
SDP
Catalog No. 502985830
Encompass_Preferred
A Chk1 inhibitor (Ki = 0.9 nM); IC50 values of 2/M checkpoint in HeLa cells; induces apoptosis in gastric cancer cell lines
Catalog No. 50-298-5830 Supplier Cayman Chemical Supplier No. 2149010 MG
A Chk1 inhibitor (Ki = 0.9 nM); IC50 values of 2/M checkpoint in HeLa cells; induces apoptosis in gastric cancer cell lines