Learn More
Cayman Chemical LY2606368, Purity: ≥98%, CAS: 1234015-52-1, 25 mg
SDP
Catalog No. 502986517
Encompass_Preferred
A Chk1 inhibitor (Ki = 0.9 nM); IC50 values of 2/M checkpoint in HeLa cells; induces apoptosis in gastric cancer cell lines
Catalog No. 50-298-6517 Supplier Cayman Chemical Supplier No. 2149025 MG
A Chk1 inhibitor (Ki = 0.9 nM); IC50 values of 2/M checkpoint in HeLa cells; induces apoptosis in gastric cancer cell lines