Learn More
Cayman Chemical LY2608204, Purity: ≥98%, CAS: 1234703-40-2, 1 mg
SDP
Catalog No. 502994222
Encompass_Preferred
An activator of glucokinase (EC50 = 42 nM); stimulates glucose metabolism in INS-1E rat insulinoma cells (mean EC50 = 579 nM); decreases plasma glucose levels in a dose-dependent manner in fasted rats (30 mg/kg)
Catalog No. 50-299-4222 Supplier Cayman Chemical Supplier No. 229141 MG
An activator of glucokinase (EC50 = 42 nM); stimulates glucose metabolism in INS-1E rat insulinoma cells (mean EC50 = 579 nM); decreases plasma glucose levels in a dose-dependent manner in fasted rats (30 mg/kg)