Learn More
Cayman Chemical LY2874455, Purity: ≥98%, CAS: 1254473-64-7, 5 mg
SDP
Catalog No. 502982997
Encompass_Preferred
A pan-FGFR inhibitor (IC50s = 2.8, 2.6, 6.4, 6, and 7 nM for FGFR1, FGFR2, FGFR3, FGFR4, and VEGFR2, respectively); inhibits proliferation of KMS-11, OPM-2, SNU-16, and KATO-III cancer cells in vitro and in tumor xenografts in mice
Catalog No. 50-298-2997 Supplier Cayman Chemical Supplier No. 208435 MG
A pan-FGFR inhibitor (IC50s = 2.8, 2.6, 6.4, 6, and 7 nM for FGFR1, FGFR2, FGFR3, FGFR4, and VEGFR2, respectively); inhibits proliferation of KMS-11, OPM-2, SNU-16, and KATO-III cancer cells in vitro and in tumor xenografts in mice