Learn More
Cayman Chemical ANTIBODY LysoFP-NO2 25mg
SDP
Catalog No. 502941963
Encompass_Preferred
A turn-on lysosome-targeted fluorescent probe for CO; transformed into the highly fluorescent derivative lysoFP-NH2 in the presence of lysosomal CO; selective for CO over reactive nitrogen, oxygen, and sulfur species; ex/em = 440/528 nm, respectively; not cytotoxic to HepG2 cells for up to five hours at 30 µM
This item is not returnable. View return policy
Catalog No. 50-294-1963 Supplier Cayman Chemical Supplier No. 2702425 MG
A turn-on lysosome-targeted fluorescent probe for CO; transformed into the highly fluorescent derivative lysoFP-NH2 in the presence of lysosomal CO; selective for CO over reactive nitrogen, oxygen, and sulfur species; ex/em = 440/528 nm, respectively; not cytotoxic to HepG2 cells for up to five hours at 30 µM