Learn More
Cayman Chemical LysoFP-NO2, Purity: ≥98%, CAS: 69408-75-9, 5 mg
SDP
Catalog No. 502939337
Encompass_Preferred
A turn-on lysosome-targeted fluorescent probe for CO; transformed into the highly fluorescent derivative lysoFP-NH2 in the presence of lysosomal CO; selective for CO over reactive nitrogen, oxygen, and sulfur species; ex/em = 440/528 nm, respectively; not cytotoxic to HepG2 cells for up to five hours at 30 µM
Catalog No. 50-293-9337 Supplier Cayman Chemical Supplier No. 270245 MG
A turn-on lysosome-targeted fluorescent probe for CO; transformed into the highly fluorescent derivative lysoFP-NH2 in the presence of lysosomal CO; selective for CO over reactive nitrogen, oxygen, and sulfur species; ex/em = 440/528 nm, respectively; not cytotoxic to HepG2 cells for up to five hours at 30 µM