Learn More
Cayman Chemical M-144, Purity: ≥98%, CAS: 2180924-19-8, 1 mg
SDP
Catalog No. 502972185
Encompass_Preferred
Differs from UR-144 by having a fluorine atom on the terminal carbon of the alkyl group as well as a methyl group attached to the central indole structure; intended for research and forensic applications
Catalog No. 50-297-2185 Supplier Cayman Chemical Supplier No. 160841 MG
Differs from UR-144 by having a fluorine atom on the terminal carbon of the alkyl group as well as a methyl group attached to the central indole structure; intended for research and forensic applications