Learn More
Cayman Chemical MM-206, Purity: ≥98%, CAS: 1809581-87-0, 10 mg
SDP
Catalog No. 502987909
Encompass_Preferred
A STAT3 inhibitor (IC50 = 1.16 µM); inhibits G-CSF-induced STAT3 phosphorylation in HL-60, Kasumi-1, and MOLM-13 AML cells (IC50s = 0.8, 1.9, and 1.1 µM, respectively); slows disease progression and improves survival in an AML mouse xenograft model (30 mg/kg per day)
Catalog No. 50-298-7909 Supplier Cayman Chemical Supplier No. 2203210 MG
A STAT3 inhibitor (IC50 = 1.16 µM); inhibits G-CSF-induced STAT3 phosphorylation in HL-60, Kasumi-1, and MOLM-13 AML cells (IC50s = 0.8, 1.9, and 1.1 µM, respectively); slows disease progression and improves survival in an AML mouse xenograft model (30 mg/kg per day)