Learn More
Cayman Chemical M2I-1, Purity: ≥98%, CAS: 312271-03-7, 5 mg
SDP
Catalog No. 502964277
Encompass_Preferred
An inhibitor of the Mad2-Cdc20 interaction; inhibits Mad2 binding to Cdc20111-138 from 6.25-100 μM; reduces paclitaxel-induced increases in mitotic duration in HeLa cells at 25 μM; reduces 4-cell embryo and blastocyst formation in hydrogen peroxide-exposed mouse zygotes from 20-120 μM, as well as increases the rate of sex chromosome mosaicism in male mouse in vitro fertilization-derived embryos
Catalog No. 50-296-4277 Supplier Cayman Chemical Supplier No. 222975 MG
An inhibitor of the Mad2-Cdc20 interaction; inhibits Mad2 binding to Cdc20111-138 from 6.25-100 μM; reduces paclitaxel-induced increases in mitotic duration in HeLa cells at 25 μM; reduces 4-cell embryo and blastocyst formation in hydrogen peroxide-exposed mouse zygotes from 20-120 μM, as well as increases the rate of sex chromosome mosaicism in male mouse in vitro fertilization-derived embryos