Learn More
Cayman Chemical M50054, Purity: ≥98%, CAS: 54135-60-3, 25 mg
SDP
Catalog No. 502980879
Encompass_Preferred
A cell-permeable inhibitor of caspase-3 activation; blocks apoptosis in a variety of cell lines induced by different stimuli, including Fas ligand and etoposide
Catalog No. 50-298-0879 Supplier Cayman Chemical Supplier No. 2081025 MG
A cell-permeable inhibitor of caspase-3 activation; blocks apoptosis in a variety of cell lines induced by different stimuli, including Fas ligand and etoposide