Learn More
Cayman Chemical Maribavir, Purity: ≥98%, CAS: 176161-24-3, 5 mg
SDP
Catalog No. 502918438
Encompass_Preferred
An antiviral agent; inhibits human CMV pUL97 protein kinase (IC50 = 3 nM); inhibits the replication of clinical isolates of CMV, including those resistant to ganciclovir, in infected MRC-5 fibroblasts (IC50s = 0.03-0.13 µM); decreases CMV titers in a mouse model of ocular CMV infection following implantation of human fetal retinal tissue at 25 or 75 mg/kg, p.o.
Catalog No. 50-291-8438 Supplier Cayman Chemical Supplier No. 396505 MG
An antiviral agent; inhibits human CMV pUL97 protein kinase (IC50 = 3 nM); inhibits the replication of clinical isolates of CMV, including those resistant to ganciclovir, in infected MRC-5 fibroblasts (IC50s = 0.03-0.13 µM); decreases CMV titers in a mouse model of ocular CMV infection following implantation of human fetal retinal tissue at 25 or 75 mg/kg, p.o.