Learn More
Cayman Chemical ANTIBODY mB-ICA 5mg
SDP
Catalog No. 502945602
Encompass_Preferred
An analytical reference standard categorized as a precursor in the synthesis of 5Cl-MMB-PICA, 5Br-MMB-PICA, MMB022, and MMB-4en-PICA butanoic acid metabolite; a metabolite of MMB-CHMICA; intended for research and forensic applications
Catalog No. 50-294-5602 Supplier Cayman Chemical Supplier No. 399515 MG
An analytical reference standard categorized as a precursor in the synthesis of 5Cl-MMB-PICA, 5Br-MMB-PICA, MMB022, and MMB-4en-PICA butanoic acid metabolite; a metabolite of MMB-CHMICA; intended for research and forensic applications