Learn More
Cayman Chemical MCB-613, Purity: ≥95%, CAS: 296792-62-6, 10 mg
SDP
Catalog No. 502967101
Encompass_Preferred
A small molecule stimulator of SRC-1, SRC-2, and SRC-3 that selectively induces paraptosis in a variety of cancer cells in vitro and reduces tumor growth in a xenograft mouse model; can increase the interaction of SRCs with the coactivators CBP and CARM1 and can induce ER stress coupled to the generation of reactive oxygen species
Catalog No. 50-296-7101 Supplier Cayman Chemical Supplier No. 1831510 MG
A small molecule stimulator of SRC-1, SRC-2, and SRC-3 that selectively induces paraptosis in a variety of cancer cells in vitro and reduces tumor growth in a xenograft mouse model; can increase the interaction of SRCs with the coactivators CBP and CARM1 and can induce ER stress coupled to the generation of reactive oxygen species