Learn More
Cayman Chemical ANTIBODY MCTR3 100ug
SDP
Catalog No. 502982306
Encompass_Preferred
A specialized pro-resolving mediator; accelerates tissue regeneration in planaria at a concentration of 1 and 100 nM; reduces neutrophil infiltration, shortens inflammatory resolution, and increases phagocytosis by macrophages; selectively reduces the eicosanoids PGD2, PGE2, PGF2α, and TXB2 in the inflammatory exudate in a mouse model of peritonitis
Catalog No. 50-298-2306 Supplier Cayman Chemical Supplier No. 19067100 UG
A specialized pro-resolving mediator; accelerates tissue regeneration in planaria at a concentration of 1 and 100 nM; reduces neutrophil infiltration, shortens inflammatory resolution, and increases phagocytosis by macrophages; selectively reduces the eicosanoids PGD2, PGE2, PGF2α, and TXB2 in the inflammatory exudate in a mouse model of peritonitis