Learn More
Cayman Chemical ANTIBODY MDL 29951 500ug
SDP
Catalog No. 502970916
Encompass_Preferred
An antagonist of NMDA receptors with high affinity for the glycine binding site (Ki = 140 nM) that is functional both in vitro and in vivo; also an agonist of GPR17 (EC50 values range from 7 nM to 6 µM, depending on the assay)
Catalog No. 50-297-0916 Supplier Cayman Chemical Supplier No. 16266500 UG
An antagonist of NMDA receptors with high affinity for the glycine binding site (Ki = 140 nM) that is functional both in vitro and in vivo; also an agonist of GPR17 (EC50 values range from 7 nM to 6 µM, depending on the assay)