Learn More
Cayman Chemical MMDPPO, Purity: ≥98% (mixture of isomers), CAS: 146322-08-9, 1 mg
SDP
Catalog No. 502988094
Encompass_Preferred
An analytical reference standard categorized as an amphetamine; an intermediate in the synthesis of (±)-MDA; intended for research and forensic applications
Catalog No. 50-298-8094 Supplier Cayman Chemical Supplier No. 315791 MG
An analytical reference standard categorized as an amphetamine; an intermediate in the synthesis of (±)-MDA; intended for research and forensic applications