Learn More
Cayman Chemical MEGA-9, Purity: ≥95%, CAS: 85261-19-4, 5 g
SDP
Catalog No. 502933020
Encompass_Preferred
A nonionic detergent that can be used to solubilize membrane proteins; CMC = 20 mM under no-salt conditions; CMCs = 2-17.1 mM under high and low salt conditions for a variety of salts; has been used to solubilize the melibiose transport carrier from E. coli membranes and reconstitute it into liposomes
Catalog No. 50-293-3020 Supplier Cayman Chemical Supplier No. 257025 G
A nonionic detergent that can be used to solubilize membrane proteins; CMC = 20 mM under no-salt conditions; CMCs = 2-17.1 mM under high and low salt conditions for a variety of salts; has been used to solubilize the melibiose transport carrier from E. coli membranes and reconstitute it into liposomes