Learn More
Cayman Chemical Metyrosine, Purity: ≥98%, CAS: 672-87-7, 50 mg
SDP
Catalog No. 502933228
Encompass_Preferred
An inhibitor of tyrosine hydroxylase; reduces norepinephrine and dopamine levels in rat brains when administered at a dose of 200 mg/kg; disrupts conditioned avoidance behavior in a dose-dependent manner; reduces dexamethasone- and cyclosporin-induced hypertension in rats
Catalog No. 50-293-3228 Supplier Cayman Chemical Supplier No. 2400150 MG
An inhibitor of tyrosine hydroxylase; reduces norepinephrine and dopamine levels in rat brains when administered at a dose of 200 mg/kg; disrupts conditioned avoidance behavior in a dose-dependent manner; reduces dexamethasone- and cyclosporin-induced hypertension in rats