Learn More
Cayman Chemical MMG-11, Purity: ≥95%, CAS: 313254-94-3, 25 mg
SDP
Catalog No. 502918565
Encompass_Preferred
An antagonist of TLR1/2 (IC50 = 0.9 µM in HEK-Blue cells expressing the human receptors); selective for TLR1/2 over the TLR2/6 heterodimer (IC50 = 7.4 µM in HEK-Blue cells expressing the human receptors); inhibits Pam3CKS4-induced TNF-α and IL-6 cytokine secretion in RAW 264.7 macrophages (IC50s = 3.3 and 0.2 µM, respectively); increases SERT and GSH activity, as well as decreases LDH activity and MDA levels, in M. avium-infected Caco-2 cells at 5 ng/ml
Catalog No. 50-291-8565 Supplier Cayman Chemical Supplier No. 3878725 MG
An antagonist of TLR1/2 (IC50 = 0.9 µM in HEK-Blue cells expressing the human receptors); selective for TLR1/2 over the TLR2/6 heterodimer (IC50 = 7.4 µM in HEK-Blue cells expressing the human receptors); inhibits Pam3CKS4-induced TNF-α and IL-6 cytokine secretion in RAW 264.7 macrophages (IC50s = 3.3 and 0.2 µM, respectively); increases SERT and GSH activity, as well as decreases LDH activity and MDA levels, in M. avium-infected Caco-2 cells at 5 ng/ml