Learn More
Cayman Chemical MIK665, Purity: ≥98%, CAS: 1799631-75-6, 5 mg
SDP
Catalog No. 502911743
Encompass_Preferred
An Mcl-1 inhibitor (Ki = 1.2 nM)
Catalog No. 50-291-1743 Supplier Cayman Chemical Supplier No. 364045 MG
An Mcl-1 inhibitor (Ki = 1.2 nM)