Learn More
Cayman Chemical MitoB-d15, Purity: ≥99% deuterated forms (d1-d15), 10 mg
SDP
Catalog No. 502980480
Encompass_Preferred
An internal standard for the quantification of MitoB by GC- or LC-MS
Catalog No. 50-298-0480 Supplier Cayman Chemical Supplier No. 1747010 MG
An internal standard for the quantification of MitoB by GC- or LC-MS