Learn More
Cayman Chemical MK-0812, Purity: ≥98%, CAS: 624733-88-6, 1 mg
SDP
Catalog No. 502985784
Encompass_Preferred
A CCR2 antagonist (IC50 = 5 nM for the human receptor); inhibits mammosphere formation of BT-549 breast cancer cells; increases CCL2 levels and decreases CD11b+ monocytes in mouse blood at 0.03, 0.1, and 0.3 mg/kg
Catalog No. 50-298-5784 Supplier Cayman Chemical Supplier No. 218031 MG
A CCR2 antagonist (IC50 = 5 nM for the human receptor); inhibits mammosphere formation of BT-549 breast cancer cells; increases CCL2 levels and decreases CD11b+ monocytes in mouse blood at 0.03, 0.1, and 0.3 mg/kg