Learn More
Cayman Chemical MK-0941, Purity: ≥98%, CAS: 1137916-97-2, 5 mg
SDP
Catalog No. 502936744
Encompass_Preferred
A glucokinase activator (EC50s = 0.24 and 0.065 µM at low (2.5 mM) and high (10 mM) concentrations of glucose, respectively); increases insulin secretion from isolated rat islets of Langerhans and glucose uptake by hepatocytes in vitro
Catalog No. 50-293-6744 Supplier Cayman Chemical Supplier No. 256425 MG
A glucokinase activator (EC50s = 0.24 and 0.065 µM at low (2.5 mM) and high (10 mM) concentrations of glucose, respectively); increases insulin secretion from isolated rat islets of Langerhans and glucose uptake by hepatocytes in vitro