Learn More
Cayman Chemical MK-1775, Purity: ≥98%, CAS: 955365-80-7, 10 mg
SDP
Catalog No. 502985510
Encompass_Preferred
An inhibitor of the checkpoint kinase Wee1 (IC50 = 5.2 nM) that prevents phosphorylation of Cdc2 at tryosine15, which abrogates the G2 DNA damage checkpoint; induces apoptosis in combination with several DNA damaging agents selectively in p53-deficient tumor cell lines
Catalog No. 50-298-5510 Supplier Cayman Chemical Supplier No. 2126610 MG
An inhibitor of the checkpoint kinase Wee1 (IC50 = 5.2 nM) that prevents phosphorylation of Cdc2 at tryosine15, which abrogates the G2 DNA damage checkpoint; induces apoptosis in combination with several DNA damaging agents selectively in p53-deficient tumor cell lines