Learn More
Cayman Chemical MK-3697, Purity: ≥98%, CAS: 1224846-01-8, 5 mg
SDP
Catalog No. 502911365
Encompass_Preferred
An orexin receptor 2 antagonist (Ki = 1.1 nM); selective for OX2R over OX1R (Ki = 3,600 nM); decreases active awake time and increases SWS and REM sleep in mice at 100 mg/kg; decreases active awake time and increases phase II SWS in dogs
Catalog No. 50-291-1365 Supplier Cayman Chemical Supplier No. 347185 MG
An orexin receptor 2 antagonist (Ki = 1.1 nM); selective for OX2R over OX1R (Ki = 3,600 nM); decreases active awake time and increases SWS and REM sleep in mice at 100 mg/kg; decreases active awake time and increases phase II SWS in dogs