Learn More
Cayman Chemical MK-3903, Purity: ≥98%, CAS: 1219737-12-8, 5 mg
SDP
Catalog No. 502988956
Encompass_Preferred
An AMPK activator (EC50 = 9 nM); selective for AMPK over a kinase panel at 10 µM, as well as CYP3A4 and CYP2D6 (IC50s = >50 µM) and PXR (EC50 = >30 µM); increases muscle and liver levels of phosphorylated ACC and reduces insulin resistance in DIO mice at 30 mg/kg; inhibits hepatic fatty acid synthesis in db/db mice
Catalog No. 50-298-8956 Supplier Cayman Chemical Supplier No. 296615 MG
An AMPK activator (EC50 = 9 nM); selective for AMPK over a kinase panel at 10 µM, as well as CYP3A4 and CYP2D6 (IC50s = >50 µM) and PXR (EC50 = >30 µM); increases muscle and liver levels of phosphorylated ACC and reduces insulin resistance in DIO mice at 30 mg/kg; inhibits hepatic fatty acid synthesis in db/db mice