Learn More
Cayman Chemical MK-7622, Purity: ≥98%, CAS: 1227923-29-6, 25 mg
SDP
Catalog No. 502947355
Encompass_Preferred
A positive allosteric modulator of M1 mAChRs; increases ACh-induced calcium flux in CHO cells expressing human M1 receptors (EC50 = 21 nM); reverses scopolamine-induced cognitive deficits in an object retrieval detour task in rhesus macaques at 0.3 and 1 mg/kg
Catalog No. 50-294-7355 Supplier Cayman Chemical Supplier No. 3440725 MG
A positive allosteric modulator of M1 mAChRs; increases ACh-induced calcium flux in CHO cells expressing human M1 receptors (EC50 = 21 nM); reverses scopolamine-induced cognitive deficits in an object retrieval detour task in rhesus macaques at 0.3 and 1 mg/kg