Learn More
Cayman Chemical MK-8617, Purity: ≥98%, CAS: 1187990-87-9, 25 mg
SDP
Catalog No. 502948224
Encompass_Preferred
A HIF-PH1, -2, and -3 inhibitor (IC50s = 1, 1, and 14 nM, respectively); selective for HIF-PH1, -2, and -3 over various CYPs (IC50s = >1.6 µM for all) and FIH (IC50 = 18 µM), as well as over a panel of 171 enzymes at 10 µM; increases serum levels of EPO and hemoglobin, as well as the number of circulating red blood cells, in rats
Catalog No. 50-294-8224 Supplier Cayman Chemical Supplier No. 3379025 MG
A HIF-PH1, -2, and -3 inhibitor (IC50s = 1, 1, and 14 nM, respectively); selective for HIF-PH1, -2, and -3 over various CYPs (IC50s = >1.6 µM for all) and FIH (IC50 = 18 µM), as well as over a panel of 171 enzymes at 10 µM; increases serum levels of EPO and hemoglobin, as well as the number of circulating red blood cells, in rats