Learn More
Cayman Chemical MK-886, Purity: ≥98%, CAS: 118414-82-7, 10 mg
SDP
Catalog No. 502987369
Encompass_Preferred
A potent FLAP antagonist that prevents 5-LO activation in vivo; inhibits LT biosynthesis in leukocytes with an IC50 value of 2.5 nM
Catalog No. 50-298-7369 Supplier Cayman Chemical Supplier No. 2175310 MG
A potent FLAP antagonist that prevents 5-LO activation in vivo; inhibits LT biosynthesis in leukocytes with an IC50 value of 2.5 nM