Learn More
Cayman Chemical MK6-83, Purity: ≥98%, CAS: 1062271-24-2, 5 mg
SDP
Catalog No. 502987133
Encompass_Preferred
A TRPML1 activator (EC50s = 110, 100, and 1,230 nM in wild type, F465L, and F408Δ-expressing fibroblasts, respectively); improves zinc trafficking out of lysosomes
Catalog No. 50-298-7133 Supplier Cayman Chemical Supplier No. 219445 MG
A TRPML1 activator (EC50s = 110, 100, and 1,230 nM in wild type, F465L, and F408Δ-expressing fibroblasts, respectively); improves zinc trafficking out of lysosomes