Learn More
Cayman Chemical MKT-077, Purity: ≥95%, CAS: 147366-41-4, 25 mg
SDP
Catalog No. 502972416
Encompass_Preferred
A cationic rhodacyanine dye that inhibits the Hsp70 family of chaperones; demonstrates antiproliferative activity against cancer cell lines (EC50s = 1.4-2.2 µM in vitro) through its ability to abrogate the interaction of the chaperone with the tumor suppressor p53
Catalog No. 50-297-2416 Supplier Cayman Chemical Supplier No. 1439525 MG
A cationic rhodacyanine dye that inhibits the Hsp70 family of chaperones; demonstrates antiproliferative activity against cancer cell lines (EC50s = 1.4-2.2 µM in vitro) through its ability to abrogate the interaction of the chaperone with the tumor suppressor p53