Learn More
Cayman Chemical ML-031, Purity: ≥95%, CAS: 852230-33-2, 10 mg
SDP
Catalog No. 502969673
Encompass_Preferred
One of the first indentified agonists of S1P2 demonstrating an EC50 value of 1 μM in a S1P reporter assay
Catalog No. 50-296-9673 Supplier Cayman Chemical Supplier No. 1517110 MG
One of the first indentified agonists of S1P2 demonstrating an EC50 value of 1 μM in a S1P reporter assay