Learn More
Cayman Chemical ML-145, Purity: ≥98%, CAS: 1164500-72-4, 1 mg
SDP
Catalog No. 502947413
Encompass_Preferred
An antagonist of GPR35; inhibits zaprinast-, cromolyn-, or pamoic acid-induced β-arrestin recruitment in in HEK293 cells expressing the human receptor (EC50s = 26, 22, and 8.3 nM, respectively); inhibits constitutive and agonist-mediated IP1 accumulation in cells expressing human GPR35
Catalog No. 50-294-7413 Supplier Cayman Chemical Supplier No. 335341 MG
An antagonist of GPR35; inhibits zaprinast-, cromolyn-, or pamoic acid-induced β-arrestin recruitment in in HEK293 cells expressing the human receptor (EC50s = 26, 22, and 8.3 nM, respectively); inhibits constitutive and agonist-mediated IP1 accumulation in cells expressing human GPR35