Learn More
Cayman Chemical ML-178, Purity: ≥95%, CAS: 1355026-47-9, 5 mg
SDP
Catalog No. 502980053
Encompass_Preferred
A selective S1P4 agonist (EC50 = 46.3 nM)
Catalog No. 50-298-0053 Supplier Cayman Chemical Supplier No. 185275 MG
A selective S1P4 agonist (EC50 = 46.3 nM)