Learn More
Cayman Chemical ML-179, Purity: ≥98%, CAS: 1883548-87-5, 1 mg
SDP
Catalog No. 502969573
Encompass_Preferred
A potent inverse agonist of LRH-1 (IC50 = 320 nM) with maximum efficacy of 40% repression; inactive at the related SF1 transcriptional activator; alters the expression of haptoglobin, SAA1, and SAA4, induces the death of MDA-MB-231 breast cancer cells, and inhibits the StAR promoter (IC50 = 2.12 µM)
Catalog No. 50-296-9573 Supplier Cayman Chemical Supplier No. 151801 MG
A potent inverse agonist of LRH-1 (IC50 = 320 nM) with maximum efficacy of 40% repression; inactive at the related SF1 transcriptional activator; alters the expression of haptoglobin, SAA1, and SAA4, induces the death of MDA-MB-231 breast cancer cells, and inhibits the StAR promoter (IC50 = 2.12 µM)