Learn More
Cayman Chemical ML-202, Purity: ≥98%, CAS: 1221186-52-2, 10 mg
SDP
Catalog No. 502915182
Encompass_Preferred
A PKM2 activator; induces PMK2 activation in a cell-free assay (AC50 = 73 nM)
Catalog No. 50-291-5182 Supplier Cayman Chemical Supplier No. 3732610 MG
A PKM2 activator; induces PMK2 activation in a cell-free assay (AC50 = 73 nM)