Learn More
Cayman Chemical ML-211, Purity: ≥95%, CAS: 2205032-89-7, 25 mg
SDP
Catalog No. 502980042
Encompass_Preferred
A dual inhibitor of LYPLA1 (IC50 = 17 nM) and LYPLA2 (IC50 = 30 nM); also inhibits the serine hydrolase ABHD11 (IC50 = 10 nM)
Catalog No. 50-298-0042 Supplier Cayman Chemical Supplier No. 1763025 MG
A dual inhibitor of LYPLA1 (IC50 = 17 nM) and LYPLA2 (IC50 = 30 nM); also inhibits the serine hydrolase ABHD11 (IC50 = 10 nM)