Learn More
Cayman Chemical ML-221, Purity: ≥98%, CAS: 877636-42-5, 25 mg
SDP
Catalog No. 502940260
Encompass_Preferred
An APJ receptor antagonist (IC50 = 4.8 μM); selective for APJ over AT1 (IC50 = >78 μM); antagonizes apelin 13-induced activation of APJ in cAMP and β-arrestin recruitment assays (IC50s = 0.7 and 1.75 μM, respectively); inhibits proliferation and angiogenesis in Mz-ChA-1 cholangiocarcinoma cells from 5-15 μM; reduces tumor growth in a Mz-ChA-1 mouse xenograft model at 150 μg/kg; reduces sciatic CCI-induced mechanical allodynia and heat hyperalgesia in rats at 10 μg/animal; inhibits pathological angiogenesis and enhances normal vessel recovery in retinal ischemic regions in a mouse model of oxygen-induced retinopathy
Catalog No. 50-294-0260 Supplier Cayman Chemical Supplier No. 2731325 MG
An APJ receptor antagonist (IC50 = 4.8 μM); selective for APJ over AT1 (IC50 = >78 μM); antagonizes apelin 13-induced activation of APJ in cAMP and β-arrestin recruitment assays (IC50s = 0.7 and 1.75 μM, respectively); inhibits proliferation and angiogenesis in Mz-ChA-1 cholangiocarcinoma cells from 5-15 μM; reduces tumor growth in a Mz-ChA-1 mouse xenograft model at 150 μg/kg; reduces sciatic CCI-induced mechanical allodynia and heat hyperalgesia in rats at 10 μg/animal; inhibits pathological angiogenesis and enhances normal vessel recovery in retinal ischemic regions in a mouse model of oxygen-induced retinopathy