Learn More
Cayman Chemical ML-228, Purity: ≥95%, CAS: 1357171-62-0, 5 mg
SDP
Catalog No. 502971855
Encompass_Preferred
An activator of the HIF signaling pathway, as demonstrated by HRE reporter assay (EC50 = 1.2 µM), HIF-1α nuclear translocation assay (EC50 = 1.3 µM), and increased VEGF expression (EC50 = 1.6 µM); activity is blocked by excess iron, suggesting that ML-228 can chelate iron
Catalog No. 50-297-1855 Supplier Cayman Chemical Supplier No. 145685 MG
An activator of the HIF signaling pathway, as demonstrated by HRE reporter assay (EC50 = 1.2 µM), HIF-1α nuclear translocation assay (EC50 = 1.3 µM), and increased VEGF expression (EC50 = 1.6 µM); activity is blocked by excess iron, suggesting that ML-228 can chelate iron