Learn More
Cayman Chemical ML-243, Purity: ≥98%, CAS: 1426576-80-8, 5 mg
SDP
Catalog No. 502969595
Encompass_Preferred
An inhibitor of cancer stem cells, displaying 32-fold selective inhibition of a breast cancer stem cell-like cell line (EC50 = 2.0 µM) over a control (mammary epithelial) cell line (EC50 = 64 µM)
Catalog No. 50-296-9595 Supplier Cayman Chemical Supplier No. 151895 MG
An inhibitor of cancer stem cells, displaying 32-fold selective inhibition of a breast cancer stem cell-like cell line (EC50 = 2.0 µM) over a control (mammary epithelial) cell line (EC50 = 64 µM)