Learn More
Cayman Chemical ML-298, Purity: ≥98%, CAS: 1426916-02-0, 5 mg
SDP
Catalog No. 502969167
Encompass_Preferred
A selective inhibitor of PLD2 (IC50 = 355 nM); decreases invasive migration of U87-MG cells when used at a concentration of 10 µM
Catalog No. 50-296-9167 Supplier Cayman Chemical Supplier No. 153495 MG
A selective inhibitor of PLD2 (IC50 = 355 nM); decreases invasive migration of U87-MG cells when used at a concentration of 10 µM